Learn More
DWK Life Sciences Watch Glass Dish, made from soda-lime glass, fused rim
Product Code. 15676925 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
70 mm
80 mm
90 mm
100 mm
120 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15676925 60 mm Pack of 10
11786958 100 mm Pack of 10
10669891 250 mm Each
11776958 80 mm Pack of 10
11796958 125 mm Pack of 10
15271839 40 mm Pack of 10
15281839 50 mm Pack of 10
15686925 70 mm Pack of 10
15211849 90 mm Pack of 10
15221849 120 mm Pack of 10
15231849 150 mm Pack of 10
15241849 200 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15676925 Supplier DWK Life Sciences Supplier No. 233213405
  • ONLINE EXCLUSIVE PRICE
    32.12€
    28.56€ / Pack of 10
    Save 3.56€
    11% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
  • Reduced risk of injury due to fused rim
Specifications
Autoclavable Not Autoclavable
Material Soda Lime Glass
For Use With (Application) Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass
No. per Pack 10/Pk.
Diameter (Metric) 60 mm