missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
West Nile Virus MP Polyclonal antibody specifically detects West Nile Virus MP in Virus samples. It is validated for ELISA
Tekniske data
Tekniske data
| Antigen | West Nile Virus MP |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | ELISA |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the middle of the West Nile Virus MP. The immunogen is located within amino acids 190 - 240. Amino Acid Squence: CTKTRHSRRSRRSLT |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?