Learn More
DWK Life Sciences WHEATON™ COMPLETEPAK™ Clear Kits
Product Code. 17942681 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Product Line:
OmniFlex 3G Igloo Lyo
OmniFlex 3G Serum
Ultra Pure Serum
Quantity:
104/Cs.
120/Cs.
278/Cs.
Closure Color:
Blue
Red
Unit Size:
1 set
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Product Line Quantity Closure Color Unit Size
17942681 2 mL OmniFlex 3G Igloo Lyo 278/Cs. Blue 1 set
17902591 2 mL OmniFlex 3G Igloo Lyo 278/Cs. Red 1 set
17902631 2 mL OmniFlex 3G Serum 278/Cs. Blue 1 set
17912631 2 mL OmniFlex 3G Serum 278/Cs. Red 1 set
17982621 2 mL Ultra Pure Serum 278/Cs. Blue 1 set
17972581 2 mL Ultra Pure Serum 278/Cs. Red 1 set
17932631 5 mL OmniFlex 3G Igloo Lyo 120/Cs. Blue 1 set
17972631 5 mL OmniFlex 3G Igloo Lyo 120/Cs. Red 1 set
17942631 5 mL OmniFlex 3G Serum 120/Cs. Blue 1 set
17942591 5 mL OmniFlex 3G Serum 120/Cs. Red 1 set
17618586 5 mL Ultra Pure Serum 120/Cs. Blue 1 set
17922631 5 mL Ultra Pure Serum 120/Cs. Blue 1 set
17952631 5 mL Ultra Pure Serum 120/Cs. Red 1 set
17962631 10 mL OmniFlex 3G Igloo Lyo 104/Cs. Blue 1 set
17982571 10 mL OmniFlex 3G Igloo Lyo 104/Cs. Red 1 set
17992631 10 mL OmniFlex 3G Serum 104/Cs. Blue 1 set
17902641 10 mL OmniFlex 3G Serum 104/Cs. Red 1 set
17972591 10 mL Ultra Pure Serum 104/Cs. Blue 1 set
17982631 10 mL Ultra Pure Serum 104/Cs. Red 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 17942681 Supplier DWK Life Sciences Supplier No. W991002CP
  • ONLINE EXCLUSIVE PRICE
    3286.50€
    3169.04€ / 1 sets
    Save 117.46€
    3% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Closure Color Blue
Color Clear
Material Type I Borosilicate Glass
Sterility Sterile
Volume (Metric) 2 mL
Includes Vial, Stopper, and Seal
No. per Case 278/Cs.
Product Line OmniFlex 3G Igloo Lyo
Closure Type OmniFlex 3G
Quantity 278/Cs.
Closure Size 13 mm
Autoclavable Autoclavable
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Type COMPLETEPAK Clear