Learn More
DWK LIFE SCIENCES™ WHEATON™ Deck Kit for Compact Roller System for Small Bottles
Product Code. 10411393 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
Compact small bottle roller culture apparatus
Wheaton benchtop small roller culture apparatus system
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10411393 Wheaton benchtop small roller culture apparatus system Each
10579311 Compact small bottle roller culture apparatus Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10411393 Supplier DWK LIFE SCIENCES™ Supplier No. W348930CH
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Expand cell cultivation capacity with the WHEATON™ Deck Kit for Compact Roller System for Small Bottles. Each deck holds two roller bottles (75-121mm diameter, 290mm long) and attaches quickly to the WHEATON™ Compact Roller System, which accepts up to two additional decks.
  • Attaches quickly and easily to the Wheaton Compact Roller System for Small Bottles, which will accept two additional decks to double or triple capacity
  • Each deck accommodates two roller bottles, 75 to 121mm in diameter and 290mm long
  • Compatible with glass or plastic roller bottles
  • Suitable for cell cultivation research and pilot applications
  • Kit includes assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions
Order Info
Weight: 5 lb. (2.3kg)
Specifications
Product Type Deck Kit
Color Charcoal
For Use With (Equipment) Wheaton benchtop small roller culture apparatus system
Includes Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions