Learn More
DWK Life Sciences WHEATON™ Reusable PVC Coliwasas and Accessories
Product Code. 12437311 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Extension and Connector
Length (Metric):
1220 mm
1830 mm
Length (English):
4 ft.
6 ft.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Length (Metric) Length (English) Unit Size
12437311 Extension and Connector 1830 mm 6 ft. Each
11734129 Extension and Connector 1220 mm 4 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12437311 Supplier DWK Life Sciences Supplier No. 885252
  • ONLINE EXCLUSIVE PRICE
    670.00€
    606.38€ / Pack of 1
    Save 63.62€
    9% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WHEATON™ Reusable PVC Coliwasas and Accessories are designed for composite liquid waste sampling across a variety of environmental and industrial applications. Durable PVC construction ensures reliable, repeatable sampling performance with compatible extension and connector options available.
  • Reusable PVC construction for durable, long-term composite liquid waste sampling
  • Suitable for a variety of environmental and industrial sampling applications
  • Compatible accessories include PVC shafts, extensions, and connectors to extend sampling reach
  • Designed for use with Wheaton Coliwasa sampling systems
  • Available in multiple configurations and lengths to meet varied sampling requirements
Specifications
Product Type Extension and Connector
Material PVC
Autoclavable Not Autoclavable
Includes 48 in. L cleaning brush
Capacity (English) 37.19 oz.
Capacity (Metric) 1100 mL
Diameter (English) Inner 1.37 in.
Diameter (English) Outer 1.61 in.
Diameter (Metric) Inner 3.5 cm
Diameter (Metric) Outer 4.1 cm
Length (English) 6 ft.
Length (Metric) 1830 mm