Learn More
DWK Life Sciences Wheaton™ 85mm Condenser
Product Code. 12401571 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric) Jacket:
85 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket Unit Size
12401571 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12401571 Supplier DWK Life Sciences Supplier No. 415501
  • ONLINE EXCLUSIVE PRICE
    164.00€
    154.70€ / Pack of 1
    Save 9.30€
    5% Off
Estimated Shipment: 08-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For extraction of analytes
  • 24-400 bottom screw thread
  • 13-425 side screw thread
Specifications
Certifications/Compliance USP Type I and ASTM E 438 Type I, Class A requirements
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Length (English) Jacket 3.34 in.
Length (Metric) Jacket 85 mm
Material Borosilicate Glass
Type Condenser