Learn More
DWK Life Sciences WHEATON™ Staining Jars and Accessories
Product Code. 10492172 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Coplin Staining Jar
Slide Grip
Slide Count:
5
5 Vertically, 10 Back to Back
Unit Size:
Pack of 2
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Slide Count Unit Size
10492172 Slide Grip 5 Pack of 2
10264732 Coplin Staining Jar 5 Vertically, 10 Back to Back Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10492172 Supplier DWK Life Sciences Supplier No. 900575
  • ONLINE EXCLUSIVE PRICE
    57.60€
    52.44€ / Pack of 2
    Save 5.16€
    8% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WHEATON™ Staining Jars and Accessories offer reliable solutions for histology and slide staining workflows. The soda-lime glass Coplin Staining Jar holds up to 10 slides and approximately 60mL of staining solution. The polypropylene Slide Grip enables safe, easy transfer of five slides.
  • Coplin Staining Jar manufactured from soda-lime glass for chemical resistance and durability
  • Holds five single 3 x 1 in. (75 x 25 mm) slides vertically, or ten slides back-to-back
  • Approximately 60 mL capacity with rectangular base
  • Suitable for staining slides or use as a developing chamber for thin-layer chromatography
  • Polypropylene Slide Grip allows easy and safe transfer of five slides
  • Slide Grip fits into Wheaton Coplin staining jars
Specifications
Autoclavable Not Autoclavable
Color Clear
Material Polypropylene
Type Slide Grip
For Use With (Application) Staining
For Use With (Equipment) Wheaton Coplin Staining Jar
Slide Count 5
Height (Metric) 35mm