missing translation for 'onlineSavingsMsg'
Learn More

WTAP Polyclonal Antibody, Invitrogen™

Product Code. 15683755
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15683755 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15683755 Supplier Invitrogen Supplier No. PA549827

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total WTAPv protein.

WTAP Polyclonal antibody specifically detects WTAP in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WTAP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WTAP
Gene Accession No. Q15007, Q9ER69
Gene Alias 2810408K05Rik; 9430038B09Rik; Female-lethal(2)D homolog; hFL(2)D; KIAA0105; Mum2; PNAS-132; pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); RGD1563824; Wilms tumor 1 associated protein; wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; Wilms tumour 1-associating protein; WT1-associated protein; WTAP
Gene Symbols WTAP
Host Species Rabbit
Immunogen Synthesized peptide derived from C-terminal of human WTAP.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499020, 60532, 9589
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.