missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
XCL2 Monoclonal antibody specifically detects XCL2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | XCL2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 06 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2 |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human XCL2 (Q9UBD3; Met1-Gly114). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?