Learn More
Phenomenex™ Zebron™ ZB-1HT, GC Capillary Column 30 m x 0.25 mm x 0.1 μm
Product Code. 31746000 Shop All Phenomenex Products
Change view
Click to view available options
Length (Metric):
30 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
31746000 30 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31746000 Supplier Phenomenex™ Supplier No. 7HGG01402
Save Up to 50% on Phenomenex Products
Use promo code 27955
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
GC columns that deliver low bleed, excellent retention, and long column life for precise, reproducible results.
  • Capillary Column
TRUSTED_SUSTAINABILITY
Specifications
Particle Size None
Film Thickness 0.1 μm
Length (Metric) 30 m
Packing Material Fused Silica
USP Type G27, G36, G41
Stationary Phase Proprietary
Type GC Column
Diameter (Metric) Inner 0.25 mm