missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF446 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17864530
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17864530 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17864530 Supplier Invitrogen™ Supplier No. PA5140015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QCGRGFDWKS VFVIHHRTHT SGPGVQSPGL ATGESTEKPP QGEVAFPHHP RRSLTGPRSY PCEE Highest antigen sequence indentity to the following orthologs: Mouse - 36%.

ZNF446 contains a KRAB and three C(2)H(2) zinc fingers. ZNF446 is a transcription repressor when fused to GAL4 DNA-binding domain and co-transfected with VP-16. Overexpression of ZNF446 in COS-7 cells inhibits the transcriptional activities of SRE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF446
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF446
Gene Accession No. Q9NWS9
Gene Alias FLJ20626; Zinc finger protein 446; zinc finger protein with KRAB and SCAN domains 20; ZKSCAN20; ZNF446; ZSCAN30; ZSCAN52
Gene Symbols ZNF446
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF446. Recombinant protein control fragment (Product #RP-101118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.