missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF447 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16359834
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16359834 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16359834 Supplier Invitrogen™ Supplier No. PA582876

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QFLGILPDKV RPWVVAQYPE SCKKAASLVE GLADVLEEPG MLLGSPAGSS SILSDGVYER HMDPLLLPGE LASPSQALGA GEIPAPSETP WLSPDPLFLE QRRVREAKTE EDGPANTEQK L Highest antigen sequence indentity to the following orthologs: Mouse - 54%, Rat - 54%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF447
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN18
Gene Accession No. Q8TBC5
Gene Alias EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18
Gene Symbols ZSCAN18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZSCAN18. Recombinant protein control fragment (Product #RP-89337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.