CAS RN 16985-05-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2,4'-(2-Pyridinyl-2methylene)diphenol, TRC
CAS: 16985-05-0 Molecular Formula: C18 H15 N O2 Molecular Weight (g/mol): 277.32 Synonym: 2,4'-[(Pyridin-2-yl)methylene]bisphenol,2-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenol,Bisacodyl Imp. B (EP) IUPAC Name: 2-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenol SMILES: Oc1ccc(cc1)C(c2ccccn2)c3ccccc3O