accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22891-52-7

IUPAC Name: 3,5-dihydro-2H-1,4-benzodioxepin-5-one
Synonyms: 2,3-dihydro-1,4-benzodioxepin-5-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: PYVNMBGWXYJCCL-UHFFFAOYSA-N
SMILES: O=C1OCCOC2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results