accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOCl

CAS RN 3874-54-2

IUPAC Name: 4-chloro-1-(4-fluorophenyl)butan-1-one
Synonyms: 4-chloro-1-(4-fluorophenyl)butan-1-one
Molecular Weight (g/mol): 200.64
Molecular Formula: C10H10ClFO
InChi Key: HXAOUYGZEOZTJO-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCCl
Molecular Weight (g/mol) 
  • (2)
  • (12)
Quantity 
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (8)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (12)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (8)
  • (2)
  • (3)