accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOO

CAS RN 5396-89-4

IUPAC Name: benzyl 3-oxobutanoate
Synonyms: benzyl acetoacetate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: WOFAGNLBCJWEOE-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results