CAS RN 548-62-9
Thermo Scientific Chemicals Crystal Violet
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3; C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Thermo Scientific Chemicals Crystal Violet, pure, indicator
Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer. | CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Thermo Scientific Chemicals Crystal Violet, certified
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Crystal Violet, ACS, 90+%
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3; C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Thermo Scientific Chemicals Crystal Violet, pure, high purity biological stain
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Thermo Scientific Chemicals Crystal violet, ACS reagent
CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol
Gentian Violet (~90%), TRC
CAS: 548-62-9 Molecular Formula: C25 H30 N3 . Cl Molecular Weight (g/mol): 407.98 Synonym: C.I. Basic Violet 3 (6CI,8CI),Methanaminium, N-[4-[bis[4-(dimethylamino)phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, chloride (9CI),12416 Violet,Adergon,Aizen Crystal Violet,Aizen Crystal Violet Extra Pure,Aizen Crystal Violet Powder,Aniline violet,Aniline violet pyoktanine,Atmonil,Avermin,Axuris,Badil,Basic Violet 3,Basic Violet BN,Basonyl Violet 610,C.I. 42555,Calcozine Violet 6BN,Calcozine Violet C,Chlorohexamethylene-p-fuchsin,Crystal Violet 10B,Crystal Violet 5BO,Crystal Violet 6B,Crystal Violet 6BO,Crystal Violet AO,Crystal Violet AON,Crystal Violet BP,Crystal Violet BPC,Crystal Violet Extra Pure,Crystal Violet Extra Pure APN,Crystal Violet Extra Pure APNX,Crystal Violet FN,Crystal Violet HL 2,Crystal Violet O,Crystal Violet Pure DSC,Crystal Violet Pure DSC Brilliant,Crystal Violet SS,Crystal Violet Technical,Crystal Violet USP,Crystal Violet chloride,Crystal violet,Gentersal,Gentian Violet B,Gentian violet,Gentiaverm,Genticid,Gentioletten,Hecto Violet R,Hectograph Violet SR,Hexamethyl violet,Hexamethyl-p-rosaniline chloride,Hexamethylpararosaniline chloride,Hidaco Brilliant Crystal Violet,Meroxyl,Meroxyl-Wander,Meroxylan,Meroxylan-Wander,Methyl Violet 10B,Methyl Violet 10BD,Methyl Violet 10BK,Methyl Violet 10BN,Methyl Violet 10BNS,Methyl Violet 10BO,Methyl Violet 5BNO,Methyl Violet 5BO,Methyl Violet 6B,Methyl Violet 6B (biological stain),Methylrosaniline chloride,Methylrosanilinium chloride,Mitsui Crystal Violet,Oxiuran,Oxycolor,Oxyozyl,Paper Blue R,Pararosaniline, N,N,N',N',N' ',N' '-hexamethyl-, chloride,Plastoresin Violet 5BO,Pyoktanin,Sanyo Fanal Violet R,Vermicid,Vianin,Viocid,Violet 5BO,Violet 612,Violet 6BN,Violet CP,Violet XXIII IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: [Cl-].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccc(cc3)N(C)C
Methyl Violet, Pure, C.I.42555, Indicator Grade, Fisher Chemical™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: 11750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]