missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet (~90%), TRC

Product Code. 30137978
Change view
Click to view available options
Quantity:
50 mg
1 g
50 g
Unit Size:
1g
50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137978 1 g 1g
30135127 50 g 50g
17930425 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137978 Supplier Toronto Research Chemicals Supplier No. TRCG3625001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25 H30 N3 . Cl
Molecular Weight (g/mol) 407.98
Synonym C.I. Basic Violet 3 (6CI,8CI), Methanaminium, N-[4-[bis[4-(dimethylamino)phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, chloride (9CI), 12416 Violet, Adergon, Aizen Crystal Violet, Aizen Crystal Violet Extra Pure, Aizen Crystal Violet Powder, Aniline violet, Aniline violet pyoktanine, Atmonil, Avermin, Axuris, Badil, Basic Violet 3, Basic Violet BN, Basonyl Violet 610, C.I. 42555, Calcozine Violet 6BN, Calcozine Violet C, Chlorohexamethylene-p-fuchsin, Crystal Violet 10B, Crystal Violet 5BO, Crystal Violet 6B, Crystal Violet 6BO, Crystal Violet AO, Crystal Violet AON, Crystal Violet BP, Crystal Violet BPC, Crystal Violet Extra Pure, Crystal Violet Extra Pure APN, Crystal Violet Extra Pure APNX, Crystal Violet FN, Crystal Violet HL 2, Crystal Violet O, Crystal Violet Pure DSC, Crystal Violet Pure DSC Brilliant, Crystal Violet SS, Crystal Violet Technical, Crystal Violet USP, Crystal Violet chloride, Crystal violet, Gentersal, Gentian Violet B, Gentian violet, Gentiaverm, Genticid, Gentioletten, Hecto Violet R, Hectograph Violet SR, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Hexamethylpararosaniline chloride, Hidaco Brilliant Crystal Violet, Meroxyl, Meroxyl-Wander, Meroxylan, Meroxylan-Wander, Methyl Violet 10B, Methyl Violet 10BD, Methyl Violet 10BK, Methyl Violet 10BN, Methyl Violet 10BNS, Methyl Violet 10BO, Methyl Violet 5BNO, Methyl Violet 5BO, Methyl Violet 6B, Methyl Violet 6B (biological stain), Methylrosaniline chloride, Methylrosanilinium chloride, Mitsui Crystal Violet, Oxiuran, Oxycolor, Oxyozyl, Paper Blue R, Pararosaniline, N,N,N',N',N' ',N' '-hexamethyl-, chloride, Plastoresin Violet 5BO, Pyoktanin, Sanyo Fanal Violet R, Vermicid, Vianin, Viocid, Violet 5BO, Violet 612, Violet 6BN, Violet CP, Violet XXIII
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES [Cl-].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccc(cc3)N(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1
Formula Weight 407.2128
Chemical Name or Material Crystal Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.