CAS RN 71056-96-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(6-Methoxy-2-naphthyl)acetonitrile, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-(6-Methoxy-2-naphthyl)acetonitrile, TRC
CAS: 71056-96-7 Molecular Formula: C13 H11 N O Molecular Weight (g/mol): 197.23 Synonym: 2-Naphthaleneacetonitrile, 6-methoxy-,6-Methoxy-2-naphthaleneacetonitrile,2-(6-Methoxy-2-naphthyl)acetonitrile,2-(6-Methoxy-2-naphthyl)ethanenitrile,2-Cyanomethyl-6-methoxynaphthalene,6-Methoxynaphthalen-2-ylacetonitrile IUPAC Name: 2-(6-methoxynaphthalen-2-yl)acetonitrile SMILES: COc1ccc2cc(CC#N)ccc2c1