CAS RN 7723-14-0
CAS RN 7723-14-0
Phosphorus, red, 97%, pure, powder
CAS: 7723-14-0 Molecular Formula: P Molecular Weight (g/mol): 30.97 MDL Number: MFCD00133771 InChI Key: OAICVXFJPJFONN-UHFFFAOYSA-N IUPAC Name: phosphorus SMILES: [P]
Phosphorus standard solution, for AAS, 1 mg/ml P in water
CAS: 7723-14-0 | P | 30.97 g/mol
Sulfur-Free Phosphorus Standard: P @ 10 wt% (100000 μg/g) in 75 cSt Hydrocarbon Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
P 3000: P @ 3000 mg/Kg in 30 cSt Oil (VISC30)
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur-Free Phosphorus Standard: P @ 5 wt% (50000 μg/g) in 75 cSt Hydrocarbon Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lubricating Oil Standard 13: Ca @ 0.100; P @ 0.010; S @ 0.450; Zn @ 0.250 wt% in Lubricating Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lubricating Oil Standard 2: Ca @ 0.600; P @ 0.005; S @ 0.050; Zn @ 0.080 wt% in Lubricating Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lubricating Oil Standard 4: Ca @ 0.500; P @ 0.030 S @ 0.150; Zn @ 0.070 wt% in Lubricating Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Non-Metals Standard: As, B, P, S, Se, Si, Te @ 100 μg/mL in 5% HCl, tr. HF, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Phosphorus Standard: P @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Phosphorus Standard: P @ 5000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2