accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 845-10-3

NaH3CNCH3NNOO(E/Z)

CAS RN 845-10-3

IUPAC Name: sodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Synonyms: sodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol): 291.29
Molecular Formula: C15H14N3NaO2
InChi Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES: [Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (14)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
Grade 
  • (2)
  • (3)
  • (3)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (7)
  • (3)
  • (1)
  • (14)
  • (3)
  • (1)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)