missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-hydroxyacetophenone, 97%

Product Code. p-7024714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399619 1 g 1g
11478717 5 g 5g
11488717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399619 Supplier Thermo Scientific Alfa Aesar Supplier No. A19884.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98619-07-9 | C8H7FO2 | 154.14 g/mol

2'-Fluoro-4'-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-4′-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98619-07-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142713
InChI Key ZCZIRBNZMFUCOH-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl ethanone, 4-hydroxy-2-fluoroacetophenone, 2-fluoro-4-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl-1-ethanone, 1-2-fluoro-4-hydroxyphenyl ethan-1-one, 1-2-fluoro-4-hydroxy-phenyl ethanone, ethanone, 1-2-fluoro-4-hydroxyphenyl, 4'-hydroxy-2'-fluoroacetophenone
PubChem CID 2724912
IUPAC Name 1-(2-fluoro-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1F

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 696642
Solubility Information Insoluble in water.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-4'-hydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.