missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyisobutyric acid, 99% (dry wt.), water <2%

Product Code. 11434524
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434524 25 g 25g
11444524 100 g 100g
11454524 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434524 Supplier Thermo Scientific Alfa Aesar Supplier No. A13146.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 594-61-6 | C4H8O3 | 104.11 g/mol

2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.

Solubility
Soluble in water, ether, alcohol, methanol and hot benzene.

Notes
Incompatible with oxidizing agents, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-61-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004459
InChI Key BWLBGMIXKSTLSX-UHFFFAOYSA-N
Synonym 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid
PubChem CID 11671
ChEBI CHEBI:50129
IUPAC Name 2-hydroxy-2-methylpropanoic acid
SMILES CC(C)(O)C(O)=O

Specifications

Melting Point 77°C to 80°C
Density 1.24
Boiling Point 114°C (12 mmHg)
Quantity 25 g
Beilstein 1744739
Sensitivity Hygroscopic
Solubility Information Soluble in water,ether,alcohol,methanol and hot benzene.
Formula Weight 104.11
Percent Purity 99%
Assay (dry wt.), water <2%
Chemical Name or Material 2-Hydroxyisobutyric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Hydroxyisobutyric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hydroxyisobutyricacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.