missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone, 98+%

Product Code. 11417026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417026 1 g 1g
11427026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417026 Supplier Thermo Scientific Alfa Aesar Supplier No. B25345.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2142-70-3 | C8H7IO | 246.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1I

Specifications

Density 1.746
Boiling Point 139°C to 140°C (12 mmHg)
Flash Point 103°C (218°F)
Refractive Index 1.618
Quantity 1 g
Beilstein 2325078
Sensitivity Light sensitive
Formula Weight 246.05
Percent Purity ≥98%
Chemical Name or Material 2'-Iodoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.