missing translation for 'onlineSavingsMsg'
Learn More

2-N-Butoxyethanol, Extra Pure, SLR, Fisher Chemical™

Product Code. 10203760 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10203760 500 mL Plastic Coated Glass bottle 500mL
10385070 2.5 L Plastic Coated Glass bottle 2.5 L
10002783 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10203760 Supplier Fisher Chemical Supplier No. B/4900/PB08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-N-Butoxyethanol, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
MDL Number 2884

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 171°C
Packaging Plastic Coated Glass bottle
Quantity 500 mL
Formula Weight 118.18g/mol
Vapor Pressure 0.8 hPa at 20°C
Physical Form Liquid
compliance-icons
Product Identifier
  • 2-Butoxyethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.