missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropylamine, TRC

Product Code. 30137254
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137254 100 g 100g
30138400 25 g 25g
30012136 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137254 Supplier Toronto Research Chemicals Supplier No. TRCD491975100G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5586-73-2
Molecular Formula C15 H17 N
Molecular Weight (g/mol) 211.3
Synonym 1-Amino-3,3-diphenylpropane, 3,3-Diphenyl-1-propanamine, 3,3-Diphenyl-1-propylamine, Diphenylpropylamine, IEM 2011, NSC 137832, γ-Phenylbenzenepropanamine;
IUPAC Name 3,3-diphenylpropan-1-amine
SMILES NCCC(c1ccccc1)c2ccccc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C15H17N/c16-12-11-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10,15H,11-12,16H2
Formula Weight 211.1361
Chemical Name or Material 3,3-Diphenylpropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.