missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbutyric acid, 97%

Product Code. 11306756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306756 5 g 5g
11316756 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306756 Supplier Thermo Scientific Alfa Aesar Supplier No. L04013.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1501-05-9 | C11H12O3 | 192.214 g/mol

4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1501-05-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00004411
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
Synonym 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid
PubChem CID 73914
IUPAC Name 5-oxo-5-phenylpentanoic acid
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O

Specifications

Melting Point 126°C to 128°C
Quantity 5 g
Beilstein 641947
Solubility Information Insoluble in water.
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material 4-Benzoylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.