Learn More
5'-Chloro-2'-hydroxy-3'-nitroacetophenone, 99%, Thermo Scientific™
Product Code. 11357239
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357239 25 g 25g
11347239 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11357239 Supplier Thermo Scientific Alfa Aesar Supplier No. H27417.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84942-40-5
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00191921
InChI Key IUNBIQBAYUBIFD-UHFFFAOYSA-N
Synonym 5'-chloro-2'-hydroxy-3'-nitroacetophenone, 1-5-chloro-2-hydroxy-3-nitrophenyl ethanone, 2-acetyl-6-nitro-4-chlorophenol, 2-acetyl-6-nitro-4-chloro phenol, 2-acetyl-4-chloro-6-nitrophenol, 5-chloro-3-nitro-2-hydroxyacetophenone, benzoic acid,5-chloro-2-hydroxy-3-nitro, ethanone, 1-5-chloro-2-hydroxy-3-nitrophenyl, 1-acetyl-5-chloro-2-hydroxy-3-nitrobenzene, pubchem3355
PubChem CID 688237
SMILES CC(=O)C1=CC(=CC(=C1O)[N+](=O)[O-])Cl
Specifications
Melting Point 132°C to 135°C
Quantity 25 g
Formula Weight 215.6
Percent Purity 99%
Chemical Name or Material 5'-Chloro-2'-hydroxy-3'-nitroacetophenone