missing translation for 'onlineSavingsMsg'
Learn More

Propinox Hydrochloride, TRC

Product Code. 30135766
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135766 50 mg 50mg
30005201 250 mg 250mg
30003322 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135766 Supplier Toronto Research Chemicals Supplier No. TRCP77150050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2765-97-1
Molecular Formula C21 H23 N O3 . Cl H
Molecular Weight (g/mol) 373.87
Synonym Diphenyl(2-propynyloxy)acetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride, α-Phenyl-α-(2-propynyloxy)benzeneacetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride), Sertal, β-(Dimethylamino)ethyl α,α-diphenyl-α-(propargyloxy)acetate Hydrochloride, Pargeverine Hydrochloride
IUPAC Name 2-(dimethylamino)ethyl 2,2-diphenyl-2-prop-2-ynoxyacetate;hydrochloride
SMILES Cl.CN(C)CCOC(=O)C(OCC#C)(c1ccccc1)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H23NO3.ClH/c1-4-16-25-21(18-11-7-5-8-12-18,19-13-9-6-10-14-19)20(23)24-17-15-22(2)3;/h1,5-14H,15-17H2,2-3H3;1H
Formula Weight 373.1445
Percent Purity >95%
Chemical Name or Material Propinox Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.