missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Mouse, Clone: PC3/188A, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 510.00€
Specifications
Antigen | CD3e |
---|---|
Clone | PC3/188A |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931158
|
EnquireBio
916-MSM10-P1 |
100 μg |
510.00€
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921158
|
EnquireBio
916-MSM10-P0 |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
CD3e | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
916 | |
A synthetic peptide corresponding to amino acids 156-168 of the cytoplasmic domain of human CD3-? chain | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
20kDa |
PC3/188A | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_000724 | |
CD3E | |
IgG1 | |
Protein A/G Purified | |
Cell surface and cytoplasmic | |
Jurkat cells. Tonsil and lymph node |