missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl 4-methoxyphenyl ketone, 98%

Product Code. 11445586
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445586 5 g 5g
11455586 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445586 Supplier Thermo Scientific Alfa Aesar Supplier No. B20725.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7152-03-6 | C11H12O2 | 176.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7152-03-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00001296
InChI Key YKZSVEVTRUSPOQ-UHFFFAOYSA-N
Synonym cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl
PubChem CID 81580
IUPAC Name cyclopropyl-(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C2CC2

Specifications

Melting Point 40°C to 44°C
Boiling Point 158°C to 160°C (4.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2046539
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material Cyclopropyl 4-methoxyphenyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.