missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic acid, 98%

Product Code. 11441237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441237 25 g 25g
11451237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441237 Supplier Thermo Scientific Alfa Aesar Supplier No. B24194.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 627-03-2 | C4H8O3 | 104.105 g/mol

Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-03-2
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00004310
InChI Key YZGQDNOIGFBYKF-UHFFFAOYSA-N
Synonym ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
PubChem CID 12301
IUPAC Name 2-ethoxyacetic acid
SMILES CCOCC(=O)O

Specifications

Density 1.102
Boiling Point 207°C
Flash Point 97°C (206°F)
Odor Pungent
Refractive Index 1.419
Quantity 25 g
UN Number UN3265
Beilstein 1743037
Solubility Information Miscible with water.
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material Ethoxyacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.