missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoropyruvate, 97%

Product Code. 11419037
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419037 5 g 5g
11429037 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419037 Supplier Thermo Scientific Alfa Aesar Supplier No. B20611.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13081-18-0 | C5H5F3O3 | 170.09 g/mol

Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and alkalis (bases). Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13081-18-0
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.09
MDL Number MFCD00114935
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
PubChem CID 2737239
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
SMILES CCOC(=O)C(=O)C(F)(F)F

Specifications

Density 1.4
Boiling Point 95°C to 98°C
Flash Point 27°C (80°F)
Refractive Index 1.3405
Quantity 5 g
UN Number UN3272
Beilstein 2087388
Solubility Information Miscible with dichloromethane.
Formula Weight 170.09
Percent Purity 97%
Chemical Name or Material Ethyl trifluoropyruvate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.