missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid-13C5, TRC

Product Code. 30004382
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004382 5 mg 5mg
30010070 2.5 mg 2.5mg
30008403 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004382 Supplier Toronto Research Chemicals Supplier No. TRCG5969615MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55443-55-5
Molecular Formula 13C5H9NO4
Molecular Weight (g/mol) 152.09
Synonym L-Glutamic-1,2,3,4,5-13C5 Acid
SMILES N[13C@@H]([13CH2][13CH2][13C](O)=O)[13C](O)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1
Formula Weight 152.0699
Percent Purity >95%
Chemical Name or Material L-Glutamic Acid-13C5
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.