Learn More
Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide
Detect both the free and protein-conjugated forms of biotin with Epredia™ Biotin Ab-2, Mouse Monoclonal Antibody.
134.64€ - 383.52€
Specifications
Antigen | Biotin Ab-2 |
---|---|
Clone | BTN.4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12628706
|
Epredia™
MS-1048-P1 |
200μg/mL | 500 μL |
261.00€
500µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12638706
|
Epredia™
MS-1048-P0 |
200μg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12618706
|
Epredia™
MS-1048-P |
200μg/mL | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12648706
|
Epredia™
MS-1048-P1ABX |
1mg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12658706
|
Epredia™
MS-1048-PABX |
1mg/mL | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.Host Species: Mouse
Clone: BTN.4
Isotype: IgG1/κ
Species Reactivity: All species.
Immunogen: Biotin conjugated to BSA
Recommended for:
- Western Blotting
- Confocal Microscopy
- Flow Cytometry
- Image Analysis
- In situ Nucleic Acid Hybridization
Note:
USA: RUO; Int'l: RUO
Specifications
Biotin Ab-2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Epitope Tags | |
Biotin conjugated to BSA | |
Primary |
BTN.4 | |
Monoclonal | |
Concentrated | |
RUO | |
All species | |
IgG2a κ |