missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD24 (GPI-Linked Surface Mucin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting, and flow cytometry experiments with Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-1.
134.64€ - 463.00€
Specifications
Antigen | CD24 (GPI-Linked Surface Mucin) Ab-1 |
---|---|
Clone | SN3 |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12649446
|
Epredia™
MS-1278-P |
200μg/mL | 1 mL |
463.00€
1mL |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12669446
|
Epredia™
MS-1278-P1 |
200μg/mL | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12659446
|
Epredia™
MS-1278-P0 |
200μg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12679446
|
Epredia™
MS-1278-P1ABX |
1mg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12699446
|
Epredia™
MS-1278-PABX |
1mg/mL | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.Host Species: Mouse
Clone: SN3
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
Molecular Weight: 30-70 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen only)
- Western Blotting
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD24 (GPI-Linked Surface Mucin) Ab-1 | |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Mouse | |
B-Cell | |
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line | |
Primary |
SN3 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2a κ |