Learn More
Chromogranin A Rabbit Polyclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A, Rabbit Polyclonal Antibody.
134.64€ - 410.04€
Specifications
Antigen | Chromogranin A |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12663287
|
Epredia
RB-9003-P0 |
Concentrated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12653287
|
Epredia
RB-9003-P1 |
Concentrated | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12613287
|
Epredia
RB-9003-P |
Concentrated | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12623287
|
Epredia
RB-9003-R7 |
Prediluted | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. This antibody is excellent for formalin-fixed, paraffin-embedded tissues with no special pretreatment.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Epitope: Not determined
Immunogen: Recombinant protein encoding human chromogranin A
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Chromogranin A | |
Polyclonal | |
Rabbit | |
Oncogenesis | |
Recombinant protein encoding human chromogranin A | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Non-human Primate, Porcine, Rabbit | |
IgG |