Learn More
Sodium 3-Mercapto-1-propanesulfonate (>90%), TRC
Product Code. 30428481
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428481 100 g -
30416065 25 g -
30410130 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30428481 Supplier Toronto Research Chemicals Supplier No. TRCS808783100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17636-10-1
Molecular Formula C3H7NaO3S2
Molecular Weight (g/mol) 178.21
Synonym 3-Mercapto-1-propanesulfonic Acid monosodium Salt, 3-Mercaptopropanesulfonic Acid, Sodium Salt, Asta 7100, MPS, Sodium 3-mercapto-1-propanesulfonate, Sodium 3-mercaptopropanesulfonate, Sodium 3-mercaptopropylsulfonate, Sodium gamma-mercaptopropanesulfonate
IUPAC Name sodium;3-sulfanylpropane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)CCCS
Specifications
Quantity 100 g
InChI Formula InChI=1S/C3H8O3S2.Na/c4-8(5,6)3-1-2-7;/h7H,1-3H2,(H,4,5,6);/q;+1/p-1
Formula Weight 177.9734
Chemical Name or Material Sodium 3-Mercapto-1-propanesulfonate