missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol, TRC

Product Code. 30005575
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005575 25 g 25g
30005806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005575 Supplier Toronto Research Chemicals Supplier No. TRCA60399525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27489-62-9
Molecular Formula C6 H13 N O
Molecular Weight (g/mol) 115.174
Synonym Cyclohexanol, 4-amino-, trans-, 4-trans-Aminocyclohexanol, 4-trans-Hydroxycyclohexylamine, trans-1,4-Cyclohexanolamine, trans-1-Amino-4-cyclohexanol, trans-1-Amino-4-hydroxycyclohexane, trans-4-Amino-1-cyclohexanol, trans-4-Hydroxycyclohexylamine
IUPAC Name 4-aminocyclohexanol
SMILES N[C@@H]1CC[C@@H](O)CC1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H13NO/c7-5-1-3-6(8)4-2-5/h5-6,8H,1-4,7H2/t5-,6-
Formula Weight 115.0997
Chemical Name or Material trans-4-Aminocyclohexanol
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.