24966 Merch P1 Product
Labels and Sign Makers
Labels and sign makers are designed to meet all of your safety and facility needs including laboratory identification and traceability. They are available in portable or benchtop models with integrated labeling apps and/or software. Choose best fit devices for use in the laboratory, in a facility, or in the field.
FEATURED
Brady™ M210-LAB Label Printer
The M210-LAB Portable Label Printer is an all-in-one labeler for research, academic, and clinical labs. It creates legible labels for flat, curved or highly textured surfaces that stay stuck for years.
24966 Merch P2 Product 1
FOR LOW VOLUME PRINTING
Brady™ BradyPrinter i7100
Get the job done, no matter the demands
A reliable industrial thermal transfer benchtop printer ideal for high volume printing, with Peel function and vial label applicator.
24966 Merch P2 Product 2
MORE COLOR OPTIONS
Brady™ i3300 Industrial Label Printer with Wi-Fi
No waste, more labels, less effort
An industrial label printer ideal for identification throughout your facility.
24966 Merch P2 Product 3
Selector Guide
Save Now - Exclusive Deals
Complete Your Order - Great Deals
FAQ
A label maker and a label printer both serve the purpose of creating labels, but they differ in their design, functionality, and typical use cases. A Label Maker is usually a handheld or desktop device with an integrated keyboard and display. It’s a standalone device that does not require a computer. It allows you to type and print labels directly from the device. A label maker is ideal for simple, on-the-spot labeling needs such as organizing files, office supplies, or personal items. A label printer is typically a peripheral device that connects to a computer or network. It’s often used with specialized software for more complex labeling tasks. A label printer is suitable for professional or high-volume labeling requirements, such as shipping labels, product labels, or barcode labels.
The choice between a thermal label maker and an ink label maker depends on your specific needs. A Thermal Label Makers doesn’t require ink or toners. It use heat to print. It require less maintenance since there’s fewer parts to replace and maintain. It’s generally quicker than ink-based printers and resistant to smudging and fading. However, it primarily prints in black and white (thermal transfer can print in color with special ribbons). Direct thermal labels can fade over time, especially with exposure to heat and light. An Ink Label Maker is capable of printing in multiple colors and on a wider variety of materials. However, it requires ink cartridges or toners, which can be expensive. It also has more parts to clean and replace and is generally slower than a thermal printer.
The lifespan of a label maker can vary widely based on factors such as the brand, model, frequency of use, and maintenance. Generally, a well-maintained label maker can last several years. Here are some considerations:
  • Quality and Brand: Higher-quality, reputable brands tend to offer more durable products.
  • Usage Frequency: Heavy, frequent use can shorten the lifespan compared to occasional use.
  • Maintenance: Regular cleaning and proper handling can extend the life of the device.
  • Environment: Operating in a clean, dust-free environment can also contribute to longevity.
On average, a good-quality label maker can last anywhere from 3 to 10 years.
Resources and Related Information
Mobile Printer Selection Guide
Find the mobile printer that best suits your needs.
24966 Merch P8 Doc 1
Safety and Facility Identification Printers
Compare different benchtop label printers.
Labels and Ribbons
Shop By Brand